MilliporeSigma
Are alsin and spartin novel interaction partners?
Biochemical and biophysical research communications (2012-09-18)
Gönenç Çobanoğlu, Mehmet Ozansoy, A Nazlı Başak
ABSTRACT
Mutations in ALS2 gene/alsin are associated with recessive forms of motor neuron disorders including Juvenile Amyotrophic Lateral Sclerosis (JALS), Infantile-onset Ascending Hereditary Spastic Paraplegia (IAHSP) and Juvenile Primary Lateral Sclerosis (JPLS). In this study, we show that alsin and another MND-linked protein, spartin are related to each other both at mRNA and protein levels in Neuro2a cells. We observed significant alterations in spartin expression in alsin knock-down conditions. We further found that both proteins colocalize in N2a cells and spartin isoform-a precipitates with alsin in the same protein complex. In the light of these results we suggest that alsin and spartin may interact each other physically.