MilliporeSigma
  • Nematocyst types and venom effects of Aurelia aurita and Velella velella from the Mediterranean Sea.
Nematocyst types and venom effects of Aurelia aurita and Velella velella from the Mediterranean Sea.
Toxicon : official journal of the International Society on Toxinology (2020-02-15)
Nurçin Killi, Guido Bonello, Gian Luigi Mariottini, Patrizia Pardini, Marina Pozzolini, Sibel Cengiz
ABSTRACT
Natural substances produced by venomous marine organisms are thought to be possible sources of useful compounds and new drugs having the potential to open new ways for pharmacology, nutrition and environmental applications. In this framework, cnidarians are very interesting being widely distributed and all are venomous organisms; so, a deep knowledge of their occurrence, morphology of venomous structures and of effects of venoms at cellular level is fundamental to evaluate the possible utilization of venomous compounds or extracts. In this research, the morphology and occurrence of nematocysts in two cnidarian species (Aurelia aurita, Velella velella), and the preliminary evaluation of the cytotoxicity of V. velella crude extract, of which cytotoxicity on cell cultures at present is unknown, were considered. The specimens were sampled in Güllük Bay, Southwestern coast of Turkey, and in the Gulf of Genova, Northwestern coast of Italy. Six nematocyst types (a-isorhiza, A-isorhiza, O-isorhiza, eurytele, polyspiras, birhopaloid) having different sizes, were observed in A. aurita, and two types (eurytele and stenotele) in V. velella. The crude extract from V. velella showed cytotoxic activity against cultured fibroblasts L929 at high doses, while inducing cell proliferation at low doses. The protein content in the extract increased remarkably after disruption of nematocysts.
Product Description
Millipore
L-Alanine, A non-essential amino acid.