MilliporeSigma
  • Validation of a high-performance liquid chromatography method for the determination of oxytetracycline, tetracycline, chlortetracycline and doxycycline in bovine milk and muscle.
Validation of a high-performance liquid chromatography method for the determination of oxytetracycline, tetracycline, chlortetracycline and doxycycline in bovine milk and muscle.
Journal of chromatography. A (2003-03-05)
A L Cinquina, F Longo, G Anastasi, L Giannetti, R Cozzani
ABSTRACT
High-performance liquid chromatography with diode-array detection (HPLC-DAD) was optimised and validated for the determination of tetracyclines in bovine milk and tissues. Milk and tissue samples were extracted and purified using a solid-phase extraction HLB Oasis cartridge and analysed using HPLC-DAD set at 365 nm. The analyses were carried out using the mobile phase of 0.01 M oxalic acid-acetonitrile-methanol (60:25:15, v/v/v) on a C8 column (250 x 4.6 mm I.D., 5 microm). Recoveries of tetracyclines from spiked samples at the three concentrations (0.5, 1 and 1.5) of the maximum residues limits (corresponding to 100 microg/kg for milk and the muscle) were higher than 81.1% in milk and 83.2% in muscle. The method was successfully validated for bovine milk and muscle in compliance with requirements set by draft SANCO/ 1805/ 2000 European Decision. The decision limit (CCalpha) was in the range 113.2-127.2 microg/kg and 107.7-129.9 micro/kg for all compounds in milk and muscle, respectively. The detection capability (CCbeta) was in the range 117.2-131.3 microg/kg and 114.9-133.1 microg/kg for all compounds in milk and muscle, respectively.
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%