MilliporeSigma
  • Challenges facing lithium batteries and electrical double-layer capacitors.
Challenges facing lithium batteries and electrical double-layer capacitors.
Angewandte Chemie (International ed. in English) (2012-09-12)
Nam-Soon Choi, Zonghai Chen, Stefan A Freunberger, Xiulei Ji, Yang-Kook Sun, Khalil Amine, Gleb Yushin, Linda F Nazar, Jaephil Cho, Peter G Bruce
ABSTRACT
Energy-storage technologies, including electrical double-layer capacitors and rechargeable batteries, have attracted significant attention for applications in portable electronic devices, electric vehicles, bulk electricity storage at power stations, and "load leveling" of renewable sources, such as solar energy and wind power. Transforming lithium batteries and electric double-layer capacitors requires a step change in the science underpinning these devices, including the discovery of new materials, new electrochemistry, and an increased understanding of the processes on which the devices depend. The Review will consider some of the current scientific issues underpinning lithium batteries and electric double-layer capacitors.
Sigma-Aldrich
Vanadium (V) oxide nanosheets