MilliporeSigma
  • Dehydration stress alters the mitogen-activated-protein kinase signaling and chaperone stress response in Xenopus laevis.
Dehydration stress alters the mitogen-activated-protein kinase signaling and chaperone stress response in Xenopus laevis.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2020-06-05)
Cheng-Wei Wu, Shannon N Tessier, Kenneth B Storey
ABSTRACT
In arid conditions, the African Clawed frog Xenopus laevis enters a state of estivation dormancy as an adaptive survival strategy. Under estivation, X. laevis experience severe dehydration stress as 25-35% of total body water is lost. Dehydration in X. laevis can lead to periods of hypoxia due to elevated blood viscosity that impedes tissue perfusion. To understand how X. laevis survives under such stress, we studied the regulation pattern of key mitogen-activated protein kinases (MAPK) and their downstream transcription factors, along with several heat shock proteins in the oxygen sensitive brain and heart tissue of X. laevis under dehydration stress. Our study revealed that the activation phosphorylation residues of MAPK including JNK and MSK and their downstream transcription factors c-Jun and ATF2 are significantly decreased in the heart under dehydration. Given that JNK, c-Jun, and ATF2 are known positive regulators of apoptosis, this regulatory pattern suggest that a state of pro-survival signals may be established in the dehydrated heart. In support of this, protein levels of HSP60, a pro-apoptotic mitochondrial chaperone, was also downregulated in the heart in response to dehydration stress. In the brain tissue, most proteins remain unchanged with the exception of the apoptosis regulating p53 transcription factor, which showed a significant decrease in its activating phosphorylation residue under dehydration. Overall, our study revealed that in the Xenopus brain and heart, a specific suppression pattern of MAPK, transcription factors, and HSP takes place to potentially establish a state of pro-survival under dehydration stress.
Product Description
Millipore
MILLIPLEX MAPK/SAPK Signaling 10-Plex Kit - Cell Signaling Multiplex Assay
Pricing and availability is not currently available.