MilliporeSigma
  • Relationship between IL-8 Circulating Levels and TLR2 Hepatic Expression in Women with Morbid Obesity and Nonalcoholic Steatohepatitis.
Relationship between IL-8 Circulating Levels and TLR2 Hepatic Expression in Women with Morbid Obesity and Nonalcoholic Steatohepatitis.
International journal of molecular sciences (2020-06-18)
Teresa Auguet, Laia Bertran, Jessica Binetti, Carmen Aguilar, Salomé Martínez, Fàtima Sabench, Jesús Miguel Lopez-Dupla, José Antonio Porras, David Riesco, Daniel Del Castillo, Cristóbal Richart
ABSTRACT
The progression of nonalcoholic fatty liver disease (NAFLD) to nonalcoholic steatohepatitis (NASH) is linked to systemic inflammation. Currently, two of the aspects that need further investigation are diagnosis and treatment of NASH. In this sense, the aim of this study was to assess the relationship between circulating levels of cytokines, hepatic expression of toll-like receptors (TLRs), and degrees of NAFLD, and to investigate whether these levels could serve as noninvasive biomarkers of NASH. The present study assessed plasma levels of cytokines in 29 normal-weight women and 82 women with morbid obesity (MO) (subclassified: normal liver (n = 29), simple steatosis (n = 32), and NASH (n = 21)). We used enzyme-linked immunosorbent assays (ELISAs) to quantify cytokine and TLR4 levels and RTqPCR to assess TLRs hepatic expression. IL-1β, IL-8, IL-10, TNF-α, tPAI-1, and MCP-1 levels were increased, and adiponectin levels were decreased in women with MO. IL-8 was significantly higher in MO with NASH than in NL. To sum up, high levels of IL-8 were associated with the diagnosis of NASH in a cohort of women with morbid obesity. Moreover, a positive correlation between TLR2 hepatic expression and IL-8 circulating levels was found.
Product Description
Millipore
MILLIPLEX® Human Adipokine Magnetic Bead Panel 1 - Endocrine Multiplex Assay, The Human Adipokine Panel 1, using the Luminex xMAP technology, enables the simultaneous analysis of five biomarkers in human serum or plasma samples.
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.