MilliporeSigma
  • Generation of a human induced pluripotent stem cell line (QBRIi009-A) from a patient with a heterozygous deletion of FOXA2.
Generation of a human induced pluripotent stem cell line (QBRIi009-A) from a patient with a heterozygous deletion of FOXA2.
Stem cell research (2020-01-29)
Ahmed K Elsayed, Maryam Aghadi, Gowher Ali, Sara Al-Khawaga, Khalid Hussain, Essam M Abdelalim
ABSTRACT
FOXA2 is a transcription factor, playing an important role during development. We established an induced pluripotent stem cell (iPSC) line, QBRIi009-A, using non-integrating Sendai virus from a 4-year-old boy, displaying a complex clinical phenotype. Molecular karyotyping and cytogenetics confirmed a de novo proximal 20p11.2 deletion with a reciprocal translocation between the short arm of chromosome 6 and 20. The deleted region (~969 kb) contains only one gene, FOXA2. The generated hiPSC line was fully characterized for its pluripotency and its genetic identity. This iPSC line provides a useful model to study FOXA2 role during human development and in disease pathogenesis.
Sigma-Aldrich
Alkaline Phosphatase Detection Kit, This Alkaline Phosphatase Detection Kit is a specific & sensitive tool for the phenotypic assessment of Embryonic Stem (ES) cell differentiation by the determination of AP activity.
Pricing and availability is not currently available.