MilliporeSigma
  • Effect of dietary raw and fermented sour cherry kernel (Prunus cerasus L.) on digestibility, intestinal morphology and caecal microflora in broiler chickens.
Effect of dietary raw and fermented sour cherry kernel (Prunus cerasus L.) on digestibility, intestinal morphology and caecal microflora in broiler chickens.
Poultry science (2020-05-18)
Emrah Gungor, Guray Erener
ABSTRACT
This study was conducted to investigate the effect of dietary raw sour cherry kernel (RC) or fermented sour cherry kernel (FC) on apparent digestibility, ileal morphology, and caecal microflora in broiler chickens. Raw sour cherry kernel was fermented by Aspergillus niger for 7 D. A total of 343 one-day-old Ross 308 male chicks were assigned to 7 dietary treatments consisting of 7 replicates of 7 broilers each. All birds were fed with a commercial diet or diets supplemented with 1%, 2%, or 4% RC or FC. The experimental period was 42 D. Apparent dry matter (DM), nitrogen and ash digestibilities were diminished (P < 0.05) by dietary RC inclusion, although dietary FC did not negatively affect (P > 0.05) nutrient digestibility. Dietary 1% FC increased (P < 0.01) the villus height to crypt depth ratio (VH:CD) compared with the other treatment groups, although RC4 reduced the villus height (VH, P < 0.001) and VH:CD (P < 0.01), compared with the control group. Dietary treatments had no effect (P > 0.05) on the crypt depth (CD). Birds fed 1% FC had the highest (P < 0.05) caecal Lactobacillus spp. counts among the treatment groups. Enterococcus spp. and Escherichia coli counts in cecum were not affected (P > 0.05) by dietary treatments. The results showed that the dietary inclusion of 1% FC improved ileal morphology and caecal microflora without any adverse effect on the apparent digestibility. These results indicate that FC has the potential to be a feed additive which improves intestinal health for broiler diets.
Product Description
Millipore
GranuCult® prime MRS agar (de MAN, ROGOSA and SHARPE), according to ISO 15214, for Lactobacillus spp., suitable for microbiology