MilliporeSigma
  • Exercise Intensity and Recovery on Circulating Brain-derived Neurotrophic Factor.
Exercise Intensity and Recovery on Circulating Brain-derived Neurotrophic Factor.
Medicine and science in sports and exercise (2019-12-10)
Joshua T Reycraft, Hashim Islam, Logan K Townsend, Grant C Hayward, Tom J Hazell, Rebecca E K Macpherson
ABSTRACT
Brain-derived neurotrophic factor (BDNF) is an exercise-induced neurotropin mediating neuroprotection and synaptic plasticity. Although exercise intensity is implicated as a potentially important mediator of BNDF release after exercise, the optimal exercise stimulus (interval vs continuous) and intensity (submaximal vs supramaximal) for augmenting circulating BDNF levels remains unknown. Irisin, an exercise-driven myokine, may also contribute to neuroprotection by upregulating BDNF. To examine the response and recovery of plasma BDNF and irisin after acute exercise of differing intensities. Eight males (23.1 ± 3.0 yr of age; V˙O2max 51.2 ± 4.4 mL·kg·min) completed four acute exercise sessions: 1) moderate-intensity continuous training (MICT, 65% V˙O2max); 2) vigorous-intensity continuous training (VICT, 85% V˙O2max); 3) sprint interval training (SIT, "all out"); and 4) no exercise (CTRL). Blood was collected preexercise as well as immediately, 30 min, and 90 min postexercise. Plasma BDNF and irisin were assessed with commercially available enzyme-linked immunosorbent assay kits. Plasma BDNF levels increased immediately after exercise in the SIT group (P < 0.0001) with plasma concentrations recovering 30 and 90 min postexercise. The BDNF levels after MICT were reduced 30 min postexercise compared with immediately postexercise (P = 0.0189), with no other changes across time points in MICT and VICT groups. Plasma BDNF area under the curve in SIT was significantly higher compared with CTRL, MICT, and VICT (P = 0.0020). No changes in plasma irisin across exercise groups and time points were found (P > 0.9999). Plasma BDNF levels increased in an intensity-dependent manner with SIT eliciting the highest BDNF concentration immediately postexercise. These results identify SIT as a time-efficient exercise modality to promote brain health through BDNF release.
Sigma-Aldrich
ChemiKine Brain Derived Neurotrophic Factor, Sandwich ELISA, This ChemiKine Brain Derived Neurotrophic Factor, Sandwich ELISA is used to measure & quantify BDNF levels in Neuroscience research.