MilliporeSigma
  • Expression of an antigen associated with basal bodies of human ciliated epithelial cells.
Expression of an antigen associated with basal bodies of human ciliated epithelial cells.
The Histochemical journal (1999-07-16)
M T Comer, M Shires, N P Goode, H J Leese, L K Trejdosiewicz, J Southgate
ABSTRACT
The process and regulation of ciliogenesis in human epithelia is little understood and many components of the cilium and associated structures have not been characterised. We have identified a monoclonal antibody, LhS28, which recognises a 44,000-45,000 M(r) protein specifically associated with human ciliated epithelial cells. Immunoperoxidase labelling of formalin-fixed paraffin wax-embedded human tissues showed that LhS28 was expressed in the sub-apical zone of ciliated epithelial cells of the Fallopian tube and upper respiratory tract, but not ciliated ependyma, non-ciliated epithelia or testis containing developing spermatozoa. Immunoelectron microscopy demonstrated that the antigen recognised by LhS28 was associated with the basal body structure of the cilium and specifically with the 9 + 0 microtubule arrays. LhS28 should be a useful tool in the identification of ciliated cells in pathological specimens and for investigating mechanisms of ciliogenesis.