MilliporeSigma
  • Spectral splitting photovoltaics using perovskite and wideband dye-sensitized solar cells.
Spectral splitting photovoltaics using perovskite and wideband dye-sensitized solar cells.
Nature communications (2015-11-06)
Takumi Kinoshita, Kazuteru Nonomura, Nam Joong Jeon, Fabrizio Giordano, Antonio Abate, Satoshi Uchida, Takaya Kubo, Sang Il Seok, Mohammad Khaja Nazeeruddin, Anders Hagfeldt, Michael Grätzel, Hiroshi Segawa
ABSTRACT
The extension of the light absorption of photovoltaics into the near-infrared region is important to increase the energy conversion efficiency. Although the progress of the lead halide perovskite solar cells is remarkable, and high conversion efficiency of >20% has been reached, their absorption limit on the long-wavelength side is ∼800 nm. To further enhance the conversion efficiency of perovskite-based photovoltaics, a hybridized system with near-infrared photovoltaics is a useful approach. Here we report a panchromatic sensitizer, coded DX3, that exhibits a broad response into the near-infrared, up to ∼1100 nm, and a photocurrent density exceeding 30 mA cm(-2) in simulated air mass 1.5 standard solar radiation. Using the DX3-based dye-sensitized solar cell in conjunction with a perovskite cell that harvests visible light, the hybridized mesoscopic photovoltaics achieved a conversion efficiency of 21.5% using a system of spectral splitting.
Sigma-Aldrich
Tin (IV) oxide nanosheets