MilliporeSigma
  • Gene expression changes induced by valproate in the process of rat hippocampal neural stem cells differentiation.
Gene expression changes induced by valproate in the process of rat hippocampal neural stem cells differentiation.
Cell biology international (2019-10-24)
Hui He, Wen Li, Beilei Shen, Heyan Zhao, Juan Liu, Jianbing Qin, Jinhong Shi, Xin Yi, Min Peng, Ran Huo, Guohua Jin
ABSTRACT
Valproate (VPA), an effective clinical approved anti-epileptic drug and mood stabilizer, has been believed to induce neuronal differentiation at the expense of inhibiting astrocytic and oligodendrocytic differentiation. Nevertheless, the involving mechanisms of it remain unclear yet. In the present study, we explored the global gene expression changes of fetus rat hippocampal neural stem cells following VPA treatment by high-throughput microarray. We obtained 874 significantly upregulated genes and 258 obviously downregulated genes (fold change > 2 and P < 0.05). Then, we performed gene ontology and pathway analyses of these differentially expressed genes and chose several genes associated with nervous system according to gene ontology analysis to conduct expression analysis to validate the reliability of the array results as well as reveal possible mechanisms of VPA. To get a better comprehension of the differentially regulated genes by VPA, we conducted protein-protein association analysis of these genes, which offered a source for further studies. In addition, we made the overlap between the VPA-downregulated genes and the predicted target genes of VPA-upregulated microRNAs (miRNAs), which were previously demonstrated. These overlapped genes may provide a source to find functional VPA/miRNA/mRNA axes during neuronal differentiation. This study first constructed a comprehensive potential downstream gene map of VPA in the process of neuronal differentiation.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, clone 5F9, clone 5F9, Upstate®, from mouse