MilliporeSigma
  • Photocyclization of diarylethylenes with a boronate moiety: a useful synthetic tool to soluble PAH building blocks.
Photocyclization of diarylethylenes with a boronate moiety: a useful synthetic tool to soluble PAH building blocks.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2020-04-29)
Mikhail Feofanov, Arber Uka, Vladimir Akhmetov, Konstantin Amsharov
ABSTRACT
The synthesis of ten ortho-fused PAHs bearing boronic pinacol ester groups (BPin) is reported. The products are obtained via modification of Mallory photocyclization in 45-99% yields. Among them are examples of highly strained molecules such as [4]helicene derivatives with BPin substituents in the cavity. The method allows double C-C coupling and tolerates more than one BPin functionality.
Sigma-Aldrich
Bis(pinacolato)diboron, 99%
Sigma-Aldrich
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)