MilliporeSigma
  • Ultrathin Thin-Film Composite Polyamide Membranes Constructed on Hydrophilic Poly(vinyl alcohol) Decorated Support Toward Enhanced Nanofiltration Performance.
Ultrathin Thin-Film Composite Polyamide Membranes Constructed on Hydrophilic Poly(vinyl alcohol) Decorated Support Toward Enhanced Nanofiltration Performance.
Environmental science & technology (2020-04-24)
Xuewu Zhu, Xiaoxiang Cheng, Xinsheng Luo, Yatao Liu, Daliang Xu, Xiaobin Tang, Zhendong Gan, Liu Yang, Guibai Li, Heng Liang
ABSTRACT
Traditional polyamide-based interfacial polymerized nanofiltration (NF) membranes exhibit upper bound features between water permeance and salt selectivity. Breaking the limits of the permeability and rejections of these composite NF membranes are highly desirable for water desalination. Herein, a high-performance NF membrane (TFC-P) was fabricated via interfacial polymerization on the poly(vinyl alcohol) (PVA) interlayered poly(ether sulfone) (PES) ultrafiltration support. Owing to the large surface area, great hydrophilicity, and high porosity of the PES-PVA support, a highly cross-linked polyamide separating layer was formed with a thickness of 9.6 nm, which was almost 90% thinner than that of the control membrane (TFC-C). In addition, the TFC-P possessed lower ζ-potential, smaller pore size, and greater surface area compared to that of the TFC-C, achieving an ultrahigh water permeance of 31.4 L m-2 h-1 bar-1 and a 99.4% Na2SO4 rejection. Importantly, the PVA interlayer strategy was further applied to a pilot NF production line and the fabricated membranes presented stable water flux and salt rejections as comparable to the lab-scaled membranes. The outstanding properties of the PVA-interlayered NF membranes highlight the feasibility of the fabrication method for practical applications, which provides a new avenue to develop robust polyamide-based NF desalination membranes for environmental water treatment.
Product Description
Millipore
Universal Microbial Limit Test Membrane, white colored, sterile, diameter 70 mm, suitable for microbiology