MilliporeSigma
  • Aquatic toxicity and mode of action of CdS and ZnS nanoparticles in four microalgae species.
Aquatic toxicity and mode of action of CdS and ZnS nanoparticles in four microalgae species.
Environmental research (2020-04-20)
Konstantin Pikula, Neli Mintcheva, Sergei A Kulinich, Alexander Zakharenko, Zhanna Markina, Vladimir Chaika, Tatiana Orlova, Yaroslav Mezhuev, Emmanouil Kokkinakis, Aristidis Tsatsakis, Kirill Golokhvast
ABSTRACT
This study reports the differences in toxic action between cadmium sulfide (CdS) and zinc sulfide (ZnS) nanoparticles (NPs) prepared by recently developed xanthate-mediated method. The aquatic toxicity of the synthesized NPs on four marine microalgae species was explored. Growth rate, esterase activity, membrane potential, and morphological changes of microalgae cells were evaluated using flow cytometry and optical microscopy. CdS and ZnS NPs demonstrated similar level of general toxicity and growth-rate inhibition to all used microalgae species, except the red algae P. purpureum. More specifically, CdS NPs caused higher inhibition of growth rate for C. muelleri and P. purpureum, while ZnS NPs were more toxic for A. ussuriensis and H. akashiwo species. Our findings suggest that the sensitivity of different microalgae species to CdS and ZnS NPs depends on the chemical composition of NPs and their ability to interact with the components of microalgal cell-wall. The red microalga was highly resistant to ZnS NPs most likely due to the presence of phycoerythrin proteins in the outer membrane bound Zn2+ cations defending their cells from further toxic influence. The treatment with CdS NPs caused morphological changes and biochemical disorder in all tested microalgae species. The toxicity of CdS NPs is based on their higher photoactivity under visible light irradiation and lower dissociation in water, which allows them to generate more reactive oxygen species and create a higher risk of oxidative stress to aquatic organisms. The results of this study contribute to our understanding of the parameters affecting the aquatic toxicity of semiconductor NPs and provide a basis for further investigations.
Sigma-Aldrich
Potassium ethyl xanthogenate, 96%