MilliporeSigma
  • Solar-Driven Carbon Nanoreactor Coupling Gold and Platinum Nanocatalysts for Alcohol Oxidations.
Solar-Driven Carbon Nanoreactor Coupling Gold and Platinum Nanocatalysts for Alcohol Oxidations.
Small (Weinheim an der Bergstrasse, Germany) (2020-06-25)
Wenjie Yang, Jinhui Zhao, Hao Tian, Lizhuo Wang, Xinyao Wang, Sheng Ye, Jian Liu, Jun Huang
ABSTRACT
This research reports gold (Au) and platinum (Pt) nanocatalysts spatially confined in a porous carbon nanosphere as a new solar-driven carbon nanoreactor (CNR). The CNRs have confined size (≈100 nm), high specific surface area, and high thermal and electrical conductivity. The black color of CNR can improve the energy harvest efficiency of the solar irradiation to thermal energy within each nanoreactor. The localized surface plasmon resonance (LSPR) on Au nanocatalysts-induced electron oscillation causes the localized heating effect inside each CNR. Therefore, the heat will be accumulated in the confined space of CNR and transferred to reaction energy to drive the alcohol oxidation on uniformly dispersed Au and Pt nanoparticles inside the nanoreactor. The energetic electrons induced by LSPR effect on the surface of Au nanoparticles are transferred to the nearby and more active Pt surface via the conductive CNR, which strongly enhances the conversion of cinnamyl alcohol from 14% on Pt-CNR up to 100% on AuPt-CNR after a 3 h reaction. Therefore, the cooperative effect of Au and Pt nanoparticles confined in the CNRs utilized in this work can largely increase the efficiency of harvesting solar energy to drive the important chemical processes.
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
3-Aminophenol, 98%