MilliporeSigma
  • Asymmetric remote C-H borylation of aliphatic amides and esters with a modular iridium catalyst.
Asymmetric remote C-H borylation of aliphatic amides and esters with a modular iridium catalyst.
Science (New York, N.Y.) (2020-08-21)
Ronald L Reyes, Miyu Sato, Tomohiro Iwai, Kimichi Suzuki, Satoshi Maeda, Masaya Sawamura
ABSTRACT
Site selectivity and stereocontrol remain major challenges in C-H bond functionalization chemistry, especially in linear aliphatic saturated hydrocarbon scaffolds. We report the highly enantioselective and site-selective catalytic borylation of remote C(sp3)-H bonds γ to the carbonyl group in aliphatic secondary and tertiary amides and esters. A chiral C-H activation catalyst was modularly assembled from an iridium center, a chiral monophosphite ligand, an achiral urea-pyridine receptor ligand, and pinacolatoboryl groups. Quantum chemical calculations support an enzyme-like structural cavity formed by the catalyst components, which bind the substrate through multiple noncovalent interactions. Versatile synthetic utility of the enantioenriched γ-borylcarboxylic acid derivatives was demonstrated.
Sigma-Aldrich
Bis(pinacolato)diboron, 99%