MilliporeSigma
  • ACE2 activator diminazene aceturate ameliorates Alzheimer's disease-like neuropathology and rescues cognitive impairment in SAMP8 mice.
ACE2 activator diminazene aceturate ameliorates Alzheimer's disease-like neuropathology and rescues cognitive impairment in SAMP8 mice.
Aging (2020-07-24)
Rui Duan, Xiao Xue, Qiao-Quan Zhang, Si-Yu Wang, Peng-Yu Gong, Yan E, Teng Jiang, Ying-Dong Zhang
ABSTRACT
Previously, we revealed that brain Ang-(1-7) deficiency was involved in the pathogenesis of sporadic Alzheimer's disease (AD). We speculated that restoration of brain Ang-(1-7) levels might have a therapeutic effect against AD. However, the relatively short duration of biological effect limited the application of Ang-(1-7) in animal experiments. Since Ang-(1-7) is generated by its metabolic enzyme ACE2, we then tested the efficacy of an ACE2 activator diminazene aceturate (DIZE) on AD-like neuropathology and cognitive impairment in senescence-accelerated mouse prone substrain 8 (SAMP8) mice, an animal model of sporadic AD. Eight-month-old SAMP8 mice were injected intraperitoneally with vehicle or DIZE once a day for 30 consecutive days. DIZE markedly elevated brain Ang-(1-7) and MAS1 levels. Meanwhile, DIZE significantly reduced the levels of Aβ1-42, hyperphosphorylated tau and pro-inflammatory cytokines in the brain. The synaptic and neuronal losses in the brain were ameliorated by DIZE. Importantly, DIZE improved spatial cognitive functions in the Morris water maze test. In conclusion, this study demonstrates that DIZE ameliorates AD-like neuropathology and rescues cognitive impairment in SAMP8 mice. These beneficial effects of DIZE may be achieved by activating brain ACE2/Ang-(1-7)/MAS1 axis. These findings highlight brain ACE2/Ang-(1-7)/MAS1 axis as a potential target for the treatment of sporadic AD.
Sigma-Aldrich
Anti-TAU antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Tau (pThr205) antibody produced in rabbit, affinity isolated antibody