MilliporeSigma
  • The first copper(ii) complex with 1,10-phenanthroline and salubrinal with interesting biochemical properties.
The first copper(ii) complex with 1,10-phenanthroline and salubrinal with interesting biochemical properties.
Metallomics : integrated biometal science (2020-04-28)
Sebastiano Masuri, Enzo Cadoni, Maria Grazia Cabiddu, Francesco Isaia, Maria Giovanna Demuru, Lukáš Moráň, David Buček, Petr Vaňhara, Josef Havel, Tiziana Pivetta
ABSTRACT
The novel copper complex [Cu(phen)2(salubrinal)](ClO4)2 (C0SAL) has been synthesised and characterised. Copper(ii) is coordinated by salubrinal through the thionic group, as shown by the UV-Vis, IR, ESI-MS and tandem mass results, together with the theoretical calculations. The formed complex showed a DPPH radical scavenging ability higher than that of salubrinal alone. Studies on lipid oxidation inhibition showed that the C0SAL concentration, required to inhibit the enzyme, was lower than that of salubrinal. The inhibition of the enzyme could take place via allosteric modulation, as suggested by docking calculations. C0SAL showed a good cytotoxic activity on A2780 cells, 82 fold higher than that of the precursor salubrinal and 1.4 fold higher than that of [Cu(phen)2(H2O)](ClO4)2. Treatment with C0SAL in SKOV3 ovarian cancer cells induced expression of GRP-78 and DDIT3 regulators of ER-stress response. The cytotoxic effect of C0SAL was reverted in the presence of TUDCA, suggesting that C0SAL induces cell death through ER-stress. In A2780 cells treated with C0SAL γ-H2AX was accumulated, suggesting that DNA damage was also involved.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Cinnamamide, predominantly trans, 97%