MilliporeSigma
  • An Investigation of the Skin Barrier Restoring Effects of a Cream and Lotion Containing Ceramides in a Multi-vesicular Emulsion in People with Dry, Eczema-Prone, Skin: The RESTORE Study Phase 1.
An Investigation of the Skin Barrier Restoring Effects of a Cream and Lotion Containing Ceramides in a Multi-vesicular Emulsion in People with Dry, Eczema-Prone, Skin: The RESTORE Study Phase 1.
Dermatology and therapy (2020-07-17)
Simon G Danby, Paul V Andrew, Kirsty Brown, John Chittock, Linda J Kay, Michael J Cork
ABSTRACT
The replenishment of skin lipids depleted in the dry skin state is a desirable therapeutic target to restore skin moisturization; however, there is limited evidence demonstrating the success of this approach through the use of topical emollients. The purpose of this study was to provide evidence of the benefits of a cream and equivalent lotion containing skin lipids in a multi-vesicular emulsion for the management of dry skin. The hypothesis was that the test cream and test lotion could sustain skin moisturization for longer than traditional emollients by sustainably delivering skin lipids. A double-blind intra-subject vehicle-controlled single open-application test on the lower legs in people with dry, atopic dermatitis (atopic eczema)-prone, skin was conducted. There were six treatment sites, three per lower leg in each participant, which were treated with the test cream, the test lotion, three reference creams commonly prescribed in the UK and no treatment as a control. After baseline measurements of skin hydration, 100 μl of the test/reference creams was applied to each of the relevant treatment sites (random site allocation). Following treatment, measurements of skin hydration and scoring of visual dryness was conducted at timed intervals (3, 6, 12 and 24 h post-product application). The test cream and lotion both significantly increased skin hydration and reduced skin dryness for at least 24 h following a single application compared to a no treatment control site. Compared to three reference emollient creams the test cream and test lotion were the only products capable of sustaining clinically meaningful improvements in skin moisturization for 24 h. The sustained moisturization imparted by the test products reduces the need for frequent emollient application, often requiring 3-4 applications per day for traditional emollients, and should reduce the high burden of managing dry skin conditions like atopic dermatitis.
Sigma-Aldrich
3-Chloro-2-methylphenol, AldrichCPR
Sigma-Aldrich
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol, AldrichCPR