MilliporeSigma
Gain-of-Function Genetic Alterations of G9a Drive Oncogenesis.
Cancer discovery (2020-04-10)
Shinichiro Kato, Qing Yu Weng, Megan L Insco, Kevin Y Chen, Sathya Muralidhar, Joanna Pozniak, Joey Mark S Diaz, Yotam Drier, Nhu Nguyen, Jennifer A Lo, Ellen van Rooijen, Lajos V Kemeny, Yao Zhan, Yang Feng, Whitney Silkworth, C Thomas Powell, Brian B Liau, Yan Xiong, Jian Jin, Julia Newton-Bishop, Leonard I Zon, Bradley E Bernstein, David E Fisher
ABSTRACT
Epigenetic regulators, when genomically altered, may become driver oncogenes that mediate otherwise unexplained pro-oncogenic changes lacking a clear genetic stimulus, such as activation of the WNT/β-catenin pathway in melanoma. This study identifies previously unrecognized recurrent activating mutations in the G9a histone methyltransferase gene, as well as G9a genomic copy gains in approximately 26% of human melanomas, which collectively drive tumor growth and an immunologically sterile microenvironment beyond melanoma. Furthermore, the WNT pathway is identified as a key tumorigenic target of G9a gain-of-function, via suppression of the WNT antagonist DKK1. Importantly, genetic or pharmacologic suppression of mutated or amplified G9a using multiple in vitro and in vivo models demonstrates that G9a is a druggable target for therapeutic intervention in melanoma and other cancers harboring G9a genomic aberrations. SIGNIFICANCE: Oncogenic G9a abnormalities drive tumorigenesis and the "cold" immune microenvironment by activating WNT signaling through DKK1 repression. These results reveal a key druggable mechanism for tumor development and identify strategies to restore "hot" tumor immune microenvironments.This article is highlighted in the In This Issue feature, p. 890.
Sigma-Aldrich
EHMT1 human, recombinant, expressed in E. coli, ≥63% (SDS-PAGE)
Sigma-Aldrich
UNC0642, ≥98% (HPLC)
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys27) Antibody, clone 614M, rabbit monoclonal, culture supernatant, clone 614M, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®