MilliporeSigma
  • Regulative Loop between β-catenin and Protein Tyrosine Receptor Type γ in Chronic Myeloid Leukemia.
Regulative Loop between β-catenin and Protein Tyrosine Receptor Type γ in Chronic Myeloid Leukemia.
International journal of molecular sciences (2020-04-01)
Luisa Tomasello, Marzia Vezzalini, Christian Boni, Massimiliano Bonifacio, Luigi Scaffidi, Mohamed Yassin, Nader Al-Dewik, Paul Takam Kamga, Mauro Krampera, Claudio Sorio
ABSTRACT
Protein tyrosine phosphatase receptor type γ (PTPRG) is a tumor suppressor gene, down-regulated in Chronic Myeloid Leukemia (CML) cells by the hypermethylation of its promoter region. β-catenin (CTNNB1) is a critical regulator of Leukemic Stem Cells (LSC) maintenance and CML proliferation. This study aims to demonstrate the antagonistic regulation between β-catenin and PTPRG in CML cells. The specific inhibition of PTPRG increases the activation state of BCR-ABL1 and modulates the expression of the BCR-ABL1- downstream gene β-Catenin. PTPRG was found to be capable of dephosphorylating β-catenin, eventually causing its cytosolic destabilization and degradation in cells expressing PTPRG. Furthermore, we demonstrated that the increased expression of β-catenin in PTPRG-negative CML cell lines correlates with DNA (cytosine-5)-methyl transferase 1 (DNMT1) over-expression, which is responsible for PTPRG promoter hypermethylation, while its inhibition or down-regulation correlates with PTPRG re-expression. We finally confirmed the role of PTPRG in regulating BCR-ABL1 and β-catenin phosphorylation in primary human CML samples. We describe here, for the first time, the existence of a regulative loop occurring between PTPRG and β-catenin, whose reciprocal imbalance affects the proliferation kinetics of CML cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPRG
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT1