MilliporeSigma
  • The oral hypoxia-inducible factor prolyl hydroxylase inhibitor enarodustat counteracts alterations in renal energy metabolism in the early stages of diabetic kidney disease.
The oral hypoxia-inducible factor prolyl hydroxylase inhibitor enarodustat counteracts alterations in renal energy metabolism in the early stages of diabetic kidney disease.
Kidney international (2020-03-17)
Sho Hasegawa, Tetsuhiro Tanaka, Tomoyuki Saito, Kenji Fukui, Takeshi Wakashima, Etsuo A Susaki, Hiroki R Ueda, Masaomi Nangaku
ABSTRACT
Hypoxia-inducible factor (HIF) prolyl hydroxylase inhibitors, also known as HIF stabilizers, increase endogenous erythropoietin production and serve as novel therapeutic agents against anemia in chronic kidney disease. HIF induces the expression of various genes related to energy metabolism as an adaptive response to hypoxia. However, it remains obscure how the metabolic reprogramming in renal tissue by HIF stabilization affects the pathophysiology of kidney diseases. Previous studies suggest that systemic metabolic disorders such as hyperglycemia and dyslipidemia cause alterations of renal metabolism, leading to renal dysfunction including diabetic kidney disease. Here, we analyze the effects of enarodustat (JTZ-951), an oral HIF stabilizer, on renal energy metabolism in the early stages of diabetic kidney disease, using streptozotocin-induced diabetic rats and alloxan-induced diabetic mice. Transcriptome analysis revealed that enarodustat counteracts the alterations in diabetic renal metabolism. Transcriptome analysis showed that fatty acid and amino acid metabolisms were upregulated in diabetic renal tissue and downregulated by enarodustat, whereas glucose metabolism was upregulated. These symmetric changes were confirmed by metabolome analysis. Whereas glycolysis and tricarboxylic acid cycle metabolites were accumulated and amino acids reduced in renal tissue of diabetic animals, these metabolic disturbances were mitigated by enarodustat. Furthermore, enarodustat increased the glutathione to glutathione disulfide ratio and relieved oxidative stress in renal tissue of diabetic animals. Thus, HIF stabilization counteracts alterations in renal energy metabolism occurring in incipient diabetic kidney disease.
Sigma-Aldrich
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications