MilliporeSigma
  • Protective effects of autophagy and NFE2L2 on reactive oxygen species-induced pyroptosis of human nucleus pulposus cells.
Protective effects of autophagy and NFE2L2 on reactive oxygen species-induced pyroptosis of human nucleus pulposus cells.
Aging (2020-04-23)
Zhibiao Bai, Wei Liu, Danshuang He, Yiyang Wang, Weiwei Yi, Changqi Luo, Jieliang Shen, Zhenming Hu
ABSTRACT
Intervertebral disc degeneration (IDD) is characterized by the decrease of nucleus pulposus cells (NPCs). With the increase of the degree of degeneration, the reactive oxygen species (ROS) in nucleus pulposus tissue increases. Pyroptosis is a newly discovered form of cell death and its relationship with oxidative stress in NPCs remains unclear. This study was performed to investigate the mechanisms of pyroptosis of NPCs under oxidative stress. NPCs were isolated from IDD patients by surgical treatment. Pyroptosis related proteins like NLR family pyrin domain containing 3(NLRP3) and PYD and CARD domain containing (PYCARD) were detected by western blot, and membrane pore formation was observed by hochest33342/PI double staining or scanning electron microscope. The results showed that ROS induced the pyroptosis of NPCs and it depended on the expression of NLRP3 and PYCARD. The increased ROS level also increased transcription factor nuclear factor, erythroid 2 like 2 (NFE2L2, Nrf2) and the autophagy of NPCs, both of which attenuated the pyroptosis. In summary, ROS induces the pyroptosis of NPCs through the NLRP3/ PYCARD pathway, and establishes negative regulation by increasing autophagy and NFE2L2. These findings may provide a better understanding of the mechanism of IDD and potential therapeutic approaches for IDD treatment.
Sigma-Aldrich
Naphthol AS-D Chloroacetate Solution
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
ML385, ≥98% (HPLC)