MilliporeSigma
  • Role of Isocitrate Dehydrogenase 2 on DNA Hydroxymethylation in Human Airway Smooth Muscle Cells.
Role of Isocitrate Dehydrogenase 2 on DNA Hydroxymethylation in Human Airway Smooth Muscle Cells.
American journal of respiratory cell and molecular biology (2020-03-10)
Bonnie H Y Yeung, Jessie Huang, Steven S An, Julian Solway, Wayne Mitzner, Wan-Yee Tang
ABSTRACT
Global DNA hydroxymethylation mediated by the TET (ten-eleven translocation) enzyme was induced in allergen-induced airway hyperresponsiveness in mouse lung tissues and specifically in isolated airway smooth muscle (ASM) cells. TET is an α-ketoglutarate (α-KG)-dependent enzyme, and the production of α-KG is catalyzed by IDH (isocitrate dehydrogenase). However, the role of IDH in the regulation of DNA hydroxymethylation in ASM cells is unknown. In comparison with nonasthmatic cells, asthmatic ASM cells exhibited higher TET activity and IDH2 (but not IDH-1 or IDH-3) gene expression levels. We modified the expression of IDH2 in ASM cells from humans with asthma by siRNA and examined the α-KG levels, TET activity, global DNA hydroxymethylation, cell proliferation, and expression of ASM phenotypic genes. Inhibition of IDH2 in asthmatic ASM cells decreased the α-KG levels, TET activity, and global DNA hydroxymethylation, and reversed the aberrant ASM phenotypes (including decreased cell proliferation and ASM phenotypic gene expression). Specifically, asthmatic cells transfected with siRNA against IDH2 showed decreased 5hmC (5-hydroxymethylcytosine) levels at the TGFB2 (transforming growth factor-β2) promoter determined by oxidative bisulfite sequencing. Taken together, our findings reveal that IDH2 plays an important role in the epigenetic regulation of ASM phenotypic changes in asthmatic ASM cells, suggesting that IDH2 is a potential therapeutic target for reversing the abnormal phenotypes seen in asthma.
Sigma-Aldrich
MISSION® esiRNA, targeting human IDH2