MilliporeSigma
  • Pathogenic DDX3X Mutations Impair RNA Metabolism and Neurogenesis during Fetal Cortical Development.
Pathogenic DDX3X Mutations Impair RNA Metabolism and Neurogenesis during Fetal Cortical Development.
Neuron (2020-03-07)
Ashley L Lennox, Mariah L Hoye, Ruiji Jiang, Bethany L Johnson-Kerner, Lindsey A Suit, Srivats Venkataramanan, Charles J Sheehan, Fernando C Alsina, Brieana Fregeau, Kimberly A Aldinger, Ching Moey, Iryna Lobach, Alexandra Afenjar, Dusica Babovic-Vuksanovic, Stéphane Bézieau, Patrick R Blackburn, Jens Bunt, Lydie Burglen, Philippe M Campeau, Perrine Charles, Brian H Y Chung, Benjamin Cogné, Cynthia Curry, Maria Daniela D'Agostino, Nataliya Di Donato, Laurence Faivre, Delphine Héron, A Micheil Innes, Bertrand Isidor, Boris Keren, Amy Kimball, Eric W Klee, Paul Kuentz, Sébastien Küry, Dominique Martin-Coignard, Ghayda Mirzaa, Cyril Mignot, Noriko Miyake, Naomichi Matsumoto, Atsushi Fujita, Caroline Nava, Mathilde Nizon, Diana Rodriguez, Lot Snijders Blok, Christel Thauvin-Robinet, Julien Thevenon, Marie Vincent, Alban Ziegler, William Dobyns, Linda J Richards, A James Barkovich, Stephen N Floor, Debra L Silver, Elliott H Sherr
ABSTRACT
De novo germline mutations in the RNA helicase DDX3X account for 1%-3% of unexplained intellectual disability (ID) cases in females and are associated with autism, brain malformations, and epilepsy. Yet, the developmental and molecular mechanisms by which DDX3X mutations impair brain function are unknown. Here, we use human and mouse genetics and cell biological and biochemical approaches to elucidate mechanisms by which pathogenic DDX3X variants disrupt brain development. We report the largest clinical cohort to date with DDX3X mutations (n = 107), demonstrating a striking correlation between recurrent dominant missense mutations, polymicrogyria, and the most severe clinical outcomes. We show that Ddx3x controls cortical development by regulating neuron generation. Severe DDX3X missense mutations profoundly disrupt RNA helicase activity, induce ectopic RNA-protein granules in neural progenitors and neurons, and impair translation. Together, these results uncover key mechanisms underlying DDX3X syndrome and highlight aberrant RNA metabolism in the pathogenesis of neurodevelopmental disease.
Sigma-Aldrich
Anti-DDX3X antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NeuN purified Antibody, from guinea pig, purified by affinity chromatography
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Laminin Antibody, Chemicon®, from rabbit