MilliporeSigma
  • Stiripentol alleviates neuropathic pain in L5 spinal nerve-transected mice.
Stiripentol alleviates neuropathic pain in L5 spinal nerve-transected mice.
Journal of anesthesia (2020-03-20)
Atsushi Fujiwara, Kenta Nakao, Takeshi Ueno, Shinji Matsumura, Seiji Ito, Toshiaki Minami
ABSTRACT
Antiepileptic drugs are used not only for the treatment of epilepsy but also for that of neuropathic pain. However, their action mechanisms have not always been well explained. Stiripentol, an effective antiepileptic drug indicated as a therapeutic for Dravet syndrome, was recently shown to act as an inhibitor of lactate dehydrogenase in astrocytes. In this present study, we examined the effect of stiripentol on neuropathic pain in L5 spinal nerve-transected mice. We carried out behavioral tests using calibrated von Frey filaments and the immunohistochemistry of glial fibrillary acidic protein, an astrocyte marker, in L5 spinal nerve-transected mice after intrathecal administration of drugs. Like other anticonvulsants such as gabapentin and carbamazepine, stiripentol alleviated mechanical hyperalgesia induced by L5 spinal nerve transection in a dose-dependent manner, when intrathecally administered to mice 7, 14, and 28 days after L5 spinal nerve transection. Likewise, α-cyano-4-hydroxycinnamic acid, a broad inhibitor of monocarboxylate transporters, diminished mechanical hyperalgesia induced by L5 spinal nerve transection. Simultaneous administration of L-lactate negated the analgesic effect elicited by stiripentol, carbamazepine or α-cyano-4-hydroxycinnamic acid, but not that by gabapentin. None of the anticonvulsants affected the immunoreactivity of glial fibrillary acidic protein. This present study demonstrated that stiripentol was effective against neuropathic pain and suggested that the astrocyte-neuron lactate shuttle was involved in such pain.
Sigma-Aldrich
Stiripentol, ≥98% (HPLC)