MilliporeSigma
  • Adipose Tissue Hypertrophy, An Aberrant Biochemical Profile and Distinct Gene Expression in Lipedema.
Adipose Tissue Hypertrophy, An Aberrant Biochemical Profile and Distinct Gene Expression in Lipedema.
The Journal of surgical research (2020-05-15)
Gunther Felmerer, Aikaterini Stylianaki, Rene Hägerling, Anna Wang, Philipp Ströbel, Maija Hollmén, Nicole Lindenblatt, Epameinondas Gousopoulos
ABSTRACT
Lipedema is a common adipose tissue disorder affecting women, characterized by a symmetric subcutaneous adipose tissue deposition, particularly of the lower extremities. Lipedema is usually underdiagnosed, thus remaining an undertreated disease. Importantly, no histopathologic or molecular hallmarks exist to clearly diagnose the disease, which is often misinterpreted as obesity or lymphedema. The aim of the present study is to characterize in detail morphologic and molecular alterations in the adipose tissue composition of lipedema patients compared with healthy controls. Detailed histopathologic and molecular characterization was performed using lipid and cytokine quantification as well as gene expression arrays. The analysis was conducted on anatomically matched skin and fat tissue biopsies as well as fasting serum probes obtained from 10 lipedema and 11 gender and body mass index-matched control patients. Histologic evaluation of the adipose tissue showed increased intercellular fibrosis and adipocyte hypertrophy. Serum analysis showed an aberrant lipid metabolism without changes in the circulating adipokines. In an adipogenesis gene array, a distinct gene expression profile associated with macrophages was observed. Histologic assessment of the immune cell infiltrate confirmed the increased presence of macrophages, without changes in the T-cell compartment. Lipedema presents a distinguishable disease with typical tissue architecture and aberrant lipid metabolism, different to obesity or lymphedema. The differentially expressed genes and immune cell infiltration profile in lipedema patients further support these findings.
Sigma-Aldrich
Direct Red 80, Dye content 25 %
Sigma-Aldrich
Elastica van Gieson staining kit, suitable for histology, for connective tissue