MilliporeSigma
  • Enhanced Wound Healing- and Inflammasome-Associated Gene Expression in TNFAIP3-Interacting Protein 1- (TNIP1-) Deficient HaCaT Keratinocytes Parallels Reduced Reepithelialization.
Enhanced Wound Healing- and Inflammasome-Associated Gene Expression in TNFAIP3-Interacting Protein 1- (TNIP1-) Deficient HaCaT Keratinocytes Parallels Reduced Reepithelialization.
Mediators of inflammation (2020-05-08)
Rambon Shamilov, Tyler W Ackley, Brian J Aneskievich
ABSTRACT
TNIP1 protein is a widely expressed, cytoplasmic inhibitor of inflammatory signaling initiated by membrane receptors such as TLRs which recognize pathogen-associated and damage-associated molecular patterns (PAMPs and DAMPs). Keratinocyte TNIP1 deficiency sensitizes cells to PAMPs and DAMPs promoting hyperresponsive expression and secretion of cytokine markers (e.g., IL-8 and IL-6) relevant to cases of chronic inflammation, like psoriasis, where TNIP1 deficiency has been reported. Here, we examined the impact of TNIP1 deficiency on gene expression and cellular responses (migration and viability) relevant to acute inflammation as typically occurs in wound healing. Using siRNA-mediated TNIP1 expression knockdown in cultured HaCaT keratinocytes, we investigated TNIP1 deficiency effects on signaling downstream of TLR3 agonism with low-concentration poly (I:C), a representative PAMP/DAMP. The combination of TNIP1 knockdown and PAMP/DAMP signaling disrupted expression of specific keratinocyte differentiation markers (e.g., transglutaminase 1 and involucrin). These same conditions promoted synergistically increased expression of wound-associated markers (e.g., S100A8, TGFβ, and CCN2) suggesting potential benefit of increased inflammatory response from reduced TNIP1 protein. Unexpectedly, poly (I:C) challenge of TNIP1-deficient cells restricted reepithelialization and reduced cell viability. In these cells, there was not only increased expression for genes associated with inflammasome assembly (e.g., ASC, procaspase 1) but also for A20, a TNIP1 partner protein that represses cell-death signaling. Despite this possibly compensatory increase in A20 mRNA, there was a decrease in phospho-A20 protein, the form necessary for quenching inflammation. Hyperresponsiveness to poly (I:C) in TNIP1-deficient keratinocytes was in part mediated through p38 and JNK pathways. Taken together, we conclude that TNIP1 deficiency promotes enhanced expression of factors associated with promoting wound healing. However, the coupled, increased potential priming of the inflammasome and reduced compensatory activity of A20 has a net negative effect on overall cell recovery potential manifested by poor reepithelialization and viability. These findings suggest a previously unrecognized role for TNIP1 protein in limiting inflammation during successful progression through early wound healing stages.
Sigma-Aldrich
MISSION® esiRNA, targeting human TNIP1