MilliporeSigma
  • Phycobiliproteins from Arthrospira Platensis (Spirulina): A New Source of Peptides with Dipeptidyl Peptidase-IV Inhibitory Activity.
Phycobiliproteins from Arthrospira Platensis (Spirulina): A New Source of Peptides with Dipeptidyl Peptidase-IV Inhibitory Activity.
Nutrients (2020-03-22)
Yuchen Li, Gilda Aiello, Carlotta Bollati, Martina Bartolomei, Anna Arnoldi, Carmen Lammi
ABSTRACT
Arthrospira platensis (spirulina) is a cyanobacterium, which contains mainly two phycobiliproteins (PBP), i.e., C-phycocyanin (C-PC) and allophycocyanin (APC). In this study, PBP were hydrolyzed using trypsin, and the composition of the hydrolysate was characterized by HPLC-ESI-MS/MS. Furthermore, the potential anti-diabetic activity was assessed by using either biochemical or cellular techniques. Findings suggest that PBP peptides inhibit DPP-IV activity in vitro with a dose-response trend and an IC50 value falling in the range between 0.5 and 1.0 mg/mL. A lower inhibition of the DPP-IV activity expressed by Caco-2 cells was observed, which was explained by a secondary metabolic degradation exerted by the same cells.
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate