MilliporeSigma
  • ASK1 Mediates Nur77 Expression in T-Cell Receptor Mediated Thymocyte Apoptosis.
ASK1 Mediates Nur77 Expression in T-Cell Receptor Mediated Thymocyte Apoptosis.
Cells (2020-03-04)
Jianxin Huo, Shengli Xu, Kong-Peng Lam
ABSTRACT
: Apoptosis signal-regulating kinase 1 (ASK1) is a mitogen-activated protein kinase kinase kinase (MAPKKK) that activates downstream JNK and p38 mitogen-activated protein kinase (MAPK) to relay death signals into cells in response to various environmental stress. However, whether ASK1 plays a role in T cell receptor (TCR)-mediated apoptosis of thymocytes is unclear. Here, we show that ASK1 is activated upon TCR stimulation and plays an important role in TCR-mediated apoptosis of thymocytes by triggering downstream JNK and p38 signaling cascades. Mechanistically, ASK1-JNK/p38 signaling leads to the upregulation of neuron-derived clone 77 (Nur77), a critical pro-apoptotic protein involved in TCR-mediated apoptosis of thymocytes. Furthermore, we demonstrate that the activation of ASK1 is negatively modulated by Akt upon TCR stimulation. Thus, our results identify a previously unappreciated signaling mechanism involving ASK1 in TCR-mediated apoptosis of thymocytes.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)