MilliporeSigma
  • Aberrant HLA-DR expression in the conjunctival epithelium after autologous serum treatment in patients with graft-versus-host disease or Sjögren's syndrome.
Aberrant HLA-DR expression in the conjunctival epithelium after autologous serum treatment in patients with graft-versus-host disease or Sjögren's syndrome.
PloS one (2020-04-22)
Katerina Jirsova, Petra Seidler Stangova, Michalis Palos, Gabriela Mahelkova, Sarka Kalasova, Ivana Rybickova, Tor Paaske Utheim, Viera Vesela
ABSTRACT
The aim of this study was to determine the effect of autologous serum (AS) eye drops on the density of human leucocyte antigen (HLA)-DR-positive epithelial cells and Langerhans cells on the ocular surface of patients with bilateral severe dry eye disease (DED) due to graft-versus-host disease (GvHD) or Sjögren's syndrome (SS). The study was conducted on 24 patients (48 eyes). AS was applied 6-10 times daily for 3 months together with regular artificial tear therapy. HLA-DR-positive cells were detected by direct immunocytochemistry on upper bulbar conjunctiva imprints obtained before and after treatment. The application of AS drops led to a statistically significant increase in the mean density of aberrant HLA-DR-positive conjunctival epithelial cells (p < 0.05) and HLA-DR-positive Langerhans cells (p < 0.05) in the GvHD group. Aberrant HLA-DR-positive epithelial cells in the SS group were decreased non-significantly. All patients reported a significant decrease in the Ocular Surface Disease Index (p < 0.01), which indicates improvement of the patient's subjective feelings after therapy. There was an expected but non-significant decrease of aberrant HLA-DR-positive conjunctival epithelial cells in the SS group only. However, the increased density of HLA-DR-positive cells, indicating slight subclinical inflammation, does not outweigh the positive effect of AS in patients with DED from GvHD.
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
Millicell® Standing Cell Culture Inserts, pore size 0.4 μm, diam. 12 mm, transparent PTFE membrane, hydrophilic, size 24 wells, sterile