MilliporeSigma
  • The β-Carboline Harmine Induces Actin Dynamic Remodeling and Abrogates the Malignant Phenotype in Tumorigenic Cells.
The β-Carboline Harmine Induces Actin Dynamic Remodeling and Abrogates the Malignant Phenotype in Tumorigenic Cells.
Cells (2020-05-14)
Ronan Le Moigne, Frédéric Subra, Manale Karam, Christian Auclair
ABSTRACT
Numerous studies have shown that alteration of actin remodeling plays a pivotal role in the regulation of morphologic and phenotypic changes leading to malignancy. In the present study, we searched for drugs that can regulate actin polymerization and reverse the malignant phenotype in cancer cells. We developed a cell-free high-throughput screening assay for the identification of compounds that induce the actin polymerization in vitro, by fluorescence anisotropy. Then, the potential of the hit compound to restore the actin cytoskeleton and reverse the malignant phenotype was checked in EWS-Fli1-transformed fibroblasts and in B16-F10 melanoma cells. A β-carboline extracted from Peganum harmala (i.e., harmine) is identified as a stimulator of actin polymerization through a mechanism independent of actin binding and requiring intracellular factors involved in a process that regulates actin kinetics. Treatment of malignant cells with non-cytotoxic concentrations of harmine induces the recovery of a non-malignant cell morphology accompanied by reorganization of the actin cytoskeleton, rescued cell-cell adhesion, inhibition of cell motility and loss of anchorage-independent growth. In conclusion, harmine induces the reversion of the malignant phenotype by a process involving the modulation of actin dynamics and is a potential anti-tumor agent acting principally through a non-cytotoxic process.
Sigma-Aldrich
Harmine hydrochloride, ≥98% (HPLC)