MilliporeSigma
  • Stereoselective vascular effects of the (R)- and (S)-enantiomers of propranolol and atenolol.
Stereoselective vascular effects of the (R)- and (S)-enantiomers of propranolol and atenolol.
Journal of cardiovascular pharmacology (1995-02-01)
K Stoschitzky, W Lindner, W Kiowski
ABSTRACT
All beta-adrenergic antagonists have an asymmetric carbon atom, and most commercially available beta-blockers consist of (R)- and (S)-enantiomers in a fixed 1:1-ratio. The drugs are believed to be contraindicated when peripheral vascular disease exists, presumably due to unopposed alpha-adrenergic vasoconstriction. However, little is known about direct vascular effects of beta-blockers or of stereoselective effects on peripheral arteries. Therefore, we investigated the effects on forearm blood flow (FBF) of brachial artery infusions of the (R)- and (S)- enantiomers of propranolol and atenolol (2, 10, and 50 micrograms/min each) and their inhibitory effects on isoprenaline (Iso)-induced vasodilatation by forearm venous occlusion plethysmography in 12 healthy subjects. Only (R)-propranolol caused an increase in FBF (+21%, p < 0.05), whereas (S)-propranolol and (R)- and (S)-atenolol had no direct effect on peripheral arteries. Vasodilatation induced by Iso was abolished by (S)-propranolol and reduced by (R)-propranolol (-56%, p < 0.05) and (S)-atenolol (-68%, p < 0.05), whereas (R)-atenolol had no effect. Our results indicate that the optically pure (R)- and (S)-enantiomers of propranolol and atenolol do not exert direct vasoconstrictive effects. Furthermore, our results confirm that predominantly (S)-enantiomers have beta-adrenoceptor blocking effects, but they also show that neither the non-beta-blocking (R)-enantiomer of propranolol nor the (S)-enantiomer of the beta 1-selective agent atenolol is completely devoid of blocking effects on vascular beta 2-adrenoceptors.
Sigma-Aldrich
(S)-(−)-Propranolol hydrochloride, ≥98% (TLC), powder