MilliporeSigma
  • Antimicrobial Peptide GH12 Prevents Dental Caries by Regulating Dental Plaque Microbiota.
Antimicrobial Peptide GH12 Prevents Dental Caries by Regulating Dental Plaque Microbiota.
Applied and environmental microbiology (2020-05-18)
Wentao Jiang, Yufei Wang, Junyuan Luo, Xiangshu Chen, Yuhao Zeng, Xinwei Li, Zening Feng, Linglin Zhang
ABSTRACT
Due to the complex microecology and microenvironment of dental plaque, novel caries prevention strategies require modulating the microbial communities ecologically and reducing the cariogenic properties effectively. Antimicrobial peptide GH12 reduced the lactic acid production and exopolysaccharide (EPS) synthesis of a Streptococcus mutans biofilm and a three-species biofilm in vitro in previous studies. However, the anticaries effects and microecological effects of GH12 remained to be investigated in a complex biofilm model in vitro and an animal caries model in vivo In the present study, GH12 at 64 mg/liter showed the most effective inhibition of lactic acid production, EPS synthesis, pH decline, and biofilm integrity of human dental plaque-derived multispecies biofilms in vitro, and GH12 at 64 mg/liter was therefore chosen for use in subsequent in vitro and in vivo assays. When treated with 64-mg/liter GH12, the dental plaque-derived multispecies biofilms sampled from healthy volunteers maintained its microbial diversity and showed a microbial community structure similar to that of the control group. In the rat caries model with a caries-promoting diet, 64-mg/liter GH12 regulated the microbiota of dental plaque, in which the abundance of caries-associated bacteria was decreased and the abundance of commensal bacteria was increased. In addition, 64-mg/liter GH12 significantly reduced the caries scores of sulcal and smooth surface caries in all locations. In conclusion, GH12 inhibited the cariogenic properties of dental plaque without perturbing the dental plaque microbiota of healthy individuals and GH12 regulated the dysbiotic microbial ecology and arrested caries development under cariogenic conditions.IMPORTANCE The anticaries effects and microecological regulation effects of the antimicrobial peptide GH12 were evaluated systematically in vitro and in vivo GH12 inhibited the cariogenic virulence of dental plaque without overintervening in the microbial ecology of healthy individuals in vitro GH12 regulated the microbial ecology of dental plaque to a certain extent in vivo under cariogenic conditions, increased the proportion of commensal bacteria, and decreased the abundance of caries-associated bacteria. GH12 significantly suppressed the incidence and severity of dental caries in vivo This study thus describes an alternative antimicrobial therapy for dental caries.
Sigma-Aldrich
N-Acetylmuramic acid, ≥98% (TLC)