MilliporeSigma
  • Hyper-Crosslinked Polymer Nanocomposites Containing Mesoporous Silica Nanoparticles with Enhanced Adsorption Towards Polar Dyes.
Hyper-Crosslinked Polymer Nanocomposites Containing Mesoporous Silica Nanoparticles with Enhanced Adsorption Towards Polar Dyes.
Polymers (2020-06-25)
Marco Guerritore, Rachele Castaldo, Brigida Silvestri, Roberto Avolio, Mariacristina Cocca, Maria Emanuela Errico, Maurizio Avella, Gennaro Gentile, Veronica Ambrogi
ABSTRACT
The development of new styrene-based hyper-crosslinked nanocomposites (HCLN) containing mesoporous silica nanoparticles (MSN) is reported here as a new strategy to obtain functional high surface area materials with an enhanced hydrophilic character. The HCLN composition, morphology and porous structure were analyzed using a multi-technique approach. The HCLN displayed a high surface area (above 1600 m2/g) and higher microporosity than the corresponding hyper-crosslinked neat resin. The enhanced adsorption properties of the HCLN towards polar organic dyes was demonstrated through the adsorption of a reactive dye, Remazol Brilliant Blue R (RB). In particular, the HCLN containing 5phr MSN showed the highest adsorption capacity of RB.
Sigma-Aldrich
p-Divinylbenzene, 85%
Sigma-Aldrich
Vinylbenzyl chloride, purum, mixture of isomers, ≥95.0%, stabilized