MilliporeSigma
  • The X-linked trichothiodystrophy-causing gene RNF113A links the spliceosome to cell survival upon DNA damage.
The X-linked trichothiodystrophy-causing gene RNF113A links the spliceosome to cell survival upon DNA damage.
Nature communications (2020-03-11)
Kateryna Shostak, Zheshen Jiang, Benoit Charloteaux, Alice Mayer, Yvette Habraken, Lars Tharun, Sebastian Klein, Xinyi Xu, Hong Quan Duong, Andrii Vislovukh, Pierre Close, Alexandra Florin, Florian Rambow, Jean-Christophe Marine, Reinhard Büttner, Alain Chariot
ABSTRACT
Prolonged cell survival occurs through the expression of specific protein isoforms generated by alternate splicing of mRNA precursors in cancer cells. How alternate splicing regulates tumor development and resistance to targeted therapies in cancer remain poorly understood. Here we show that RNF113A, whose loss-of-function causes the X-linked trichothiodystrophy, is overexpressed in lung cancer and protects from Cisplatin-dependent cell death. RNF113A is a RNA-binding protein which regulates the splicing of multiple candidates involved in cell survival. RNF113A deficiency triggers cell death upon DNA damage through multiple mechanisms, including apoptosis via the destabilization of the prosurvival protein MCL-1, ferroptosis due to enhanced SAT1 expression, and increased production of ROS due to altered Noxa1 expression. RNF113A deficiency circumvents the resistance to Cisplatin and to BCL-2 inhibitors through the destabilization of MCL-1, which thus defines spliceosome inhibitors as a therapeutic approach to treat tumors showing acquired resistance to specific drugs due to MCL-1 stabilization.
Sigma-Aldrich
Anti-RNF113A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution