MilliporeSigma
  • D-methionine improves cisplatin-induced anorexia and dyspepsia syndrome by attenuating intestinal tryptophan hydroxylase 1 activity and increasing plasma leptin concentration.
D-methionine improves cisplatin-induced anorexia and dyspepsia syndrome by attenuating intestinal tryptophan hydroxylase 1 activity and increasing plasma leptin concentration.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2020-01-29)
Yi-Sin Wong, Meei-Yn Lin, Pei-Fen Liu, Jiunn-Liang Ko, Guan-Ting Huang, Dom-Gene Tu, Chu-Chyn Ou
ABSTRACT
Cisplatin is a widely used antineoplastic drug. However, cisplatin-induced dyspepsia syndromes, including delayed gastric emptying, gastric distension, early satiety, nausea, and vomiting, often force patients to take doses lower than those prescribed or even refuse treatment. D-methionine has an appetite-enhancing effect and alleviates weight loss during cisplatin treatment. This work established a model of anorexia and dyspepsia symptoms with intraperitoneal injection of cisplatin (5 mg/kg) once a week for three cycles. Presupplementation with or without D-methionine (300 mg/kg) was performed. Orexigenic and anorexigenic hormones (ghrelin, leptin, and glucagon-like peptide-1), tryptophan hydroxylase 1 (TPH1), 5-hydroxytryptamine receptors (5-HT2C and 5-HT3 ), and hypothalamic feeding-related peptides were measured by immunohistochemistry staining, enzyme-linked immunosorbent assay, and real-time PCR assay. Cisplatin administration caused marked decrease in appetite and body weight, promoted adipose and fat tissue atrophy, and delayed gastric emptying and gastric distension, and D-methionine preadministration prior to cisplatin administration significantly ameliorated these side effects. Besides, cisplatin induced an evident increase in serum ghrelin level, TPH1 activity, and 5-HT3 receptor expression in the intestine and decreased plasma leptin levels and gastric ghrelin mRNA gene expression levels. D-methionine supplementation recovered these changes. The expression of orexigenic neuropeptide Y/agouti-related peptide and anorexigenic cocaine- and amphetamine-regulated transcript proopiomelanocortin neurons were altered by D-methionine supplementation in cisplatin-induced anorexia rats. D-methionine supplementation prevents cisplatin-induced anorexia and dyspepsia syndrome possibly by attenuating intestinal tryptophan hydroxylase 1 activity and increasing plasma leptin concentration. Therefore, D-methionine can be used as an adjuvant therapy for treating cisplatin-induced adverse effects.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
D-Methionine, ≥98% (HPLC)