MilliporeSigma
  • Amino group decorated coordination polymers for enhanced detection of folic acid.
Amino group decorated coordination polymers for enhanced detection of folic acid.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2020-05-14)
Yan-Fang Jing, David James Young, Qin Huang, Yan Mi, Shu-Cong Zhang, Fei-Long Hu
ABSTRACT
A series of fluorescent coordination polymers (CPs) {[Cd2(CH3-bpeb)2(BDC)2] CP1, (BDC)0.5/(NH2-BDC)0.5-CP1, (BDC)0.34/(NH2-BDC)0.66-CP1, (BDC)0.25/(NH2-BDC)0.75-CP1, (BDC)0.2/(NH2-BDC)0.8-CP1, (NH2-BDC)-CP1} were prepared from conjugated ligand 4,4'-((2-methyl-1,4-phenylene)bis(ethene-2,1-diyl))bipyridine (CH3-bpeb), terephthalic acid (BDC), aminoterephthalic acid (NH2-BDC) and CdSO4 under solvothermal conditions. The fluorescence of aqueous suspensions of these CPs was quenched by folic acid (FA) in a concentration dependent manner. The efficiency of quenching increasing with an increased proportion of NH2-BDC ligand in the CP with (NH2-BDC)-CP1 exhibiting a low detection limit of 1.7 × 10-7 M.
Sigma-Aldrich
Hexaphenylbenzene, 98%
Sigma-Aldrich
TNP, ≥95% (HPLC)