MilliporeSigma
Induction of ligand promiscuity of αVβ3 integrin by mechanical force.
Journal of cell science (2020-03-21)
Michael Bachmann, Markus Schäfer, Vasyl V Mykuliak, Marta Ripamonti, Lia Heiser, Kai Weißenbruch, Sarah Krübel, Clemens M Franz, Vesa P Hytönen, Bernhard Wehrle-Haller, Martin Bastmeyer
ABSTRACT
αVβ3 integrin can bind to multiple extracellular matrix proteins, including vitronectin (Vn) and fibronectin (Fn), which are often presented to cells in culture as homogenous substrates. However, in tissues, cells experience highly complex and changing environments. To better understand integrin ligand selection in such complex environments, we employed binary-choice substrates of Fn and Vn to dissect αVβ3 integrin-mediated binding to different ligands on the subcellular scale. Super-resolution imaging revealed that αVβ3 integrin preferred binding to Vn under various conditions. In contrast, binding to Fn required higher mechanical load on αVβ3 integrin. Integrin mutations, structural analysis and chemical inhibition experiments indicated that the degree of hybrid domain swing-out is relevant for the selection between Fn and Vn; only a force-mediated, full hybrid domain swing-out facilitated αVβ3-Fn binding. Thus, force-dependent conformational changes in αVβ3 integrin increased the diversity of available ligands for binding and therefore enhanced the ligand promiscuity of this integrin.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid
Sigma-Aldrich
HEPBS, ≥99% (titration)
Sigma-Aldrich
Osteopontin from bovine milk, lyophilized powder
Sigma-Aldrich
Acrylic acid N-hydroxysuccinimide ester, ≥90% (GC)
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Vitronectin from human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Vitronectin antibody produced in mouse, clone VIT-2, ascites fluid
Sigma-Aldrich
Vitronectin human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture