MilliporeSigma
  • Secondary metabolites from Bacillus sp. MERNA97 extract attenuates the oxidative stress, genotoxicity and cytotoxicity of aflatoxin B1 in rats.
Secondary metabolites from Bacillus sp. MERNA97 extract attenuates the oxidative stress, genotoxicity and cytotoxicity of aflatoxin B1 in rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-05-19)
Mosaad A Abdel-Wahhab, Aziza A El-Nekeety, Amal S Hathout, Asmaa S Salman, Sekena H Abdel-Aziem, Nabila S Hassan, Mohamed S Abdel-Aziz
ABSTRACT
This study aimed to determine the bioactive compounds of Bacillus sp. MERNA97 extract and to evaluate their efficacy against the oxidative damage, genotoxicity, chromosomal aberration and DNA fragmentation in rats treated with AFB1. Sixty male Sprague-Dawley rats were divided into 6 groups and treated for 6 weeks and included the control group, AFB1-treated group (80 μg/kg b. w), the groups treated with Bacillus extract (BE) at low (2 mg/kg b.w) or high (4 mg/kg b.w) dose and the groups treated with AFB1 plus BE at the two doses. Blood and tissues samples were collected for different assays. The GC-MS results revealed the isolation of 44 compounds belong to different classes. The in vivo results showed that AFB1 disturbs all the biochemical parameters, oxidative stress markers, cytokines gene expression chromosomal aberration and DNA fragmentation along with the histological changes in the liver tissue. BE at the two tested doses induced a significant improvement in all parameters tested and the histological picture in a dose dependent manner. It could be concluded that the extract of Bacillus sp. MERNA97 isolated from the marine environment in the Red Sea is a promise as a source of novel compounds with therapeutically benefits.
Sigma-Aldrich
Cyclopentadecanone, 98%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)