MilliporeSigma
  • Assessing the Influence of the Sourcing Voltage on Polyaniline Composites for Stress Sensing Applications.
Assessing the Influence of the Sourcing Voltage on Polyaniline Composites for Stress Sensing Applications.
Polymers (2020-05-23)
Andrés Felipe Cruz-Pacheco, Leonel Paredes-Madrid, Jahir Orozco, Jairo Alberto Gómez-Cuaspud, Carlos R Batista-Rodríguez, Carlos Andrés Palacio Gómez
ABSTRACT
Polyaniline (PANI) has recently gained great attention due to its outstanding electrical properties and ease of processability; these characteristics make it ideal for the manufacturing of polymer blends. In this study, the processing and piezoresistive characterization of polymer composites resulting from the blend of PANI with ultra-high molecular weight polyethylene (UHMWPE) in different weight percentages (wt %) is reported. The PANI/UHMWPE composites were uniformly homogenized by mechanical mixing and the pellets were manufactured by compression molding. A total of four pellets were manufactured, with PANI percentages of 20, 25, 30 and 35 wt %. Fourier-transform infrared (FTIR) spectroscopy, thermogravimetric analysis (TGA), differential thermal analysis (DTA), scanning electron microscopy (SEM) and energy-dispersive X-ray spectroscopy (EDS) were used to confirm the effective distribution of PANI and UHMWPE particles in the pellets. A piezoresistive characterization was performed on the basis of compressive forces at different voltages; it was found that the error metrics of hysteresis and drift were influenced by the operating voltage. In general, larger voltages lowered the error metrics, but a reduction in sensor sensitivity came along with voltage increments. In an attempt to explain such a phenomenon, the authors developed a microscopic model for the piezoresistive response of PANI composites, aiming towards a broader usage of PANI composites in strain/stress sensing applications as an alternative to carbonaceous materials.
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Pricing and availability is not currently available.
Sigma-Aldrich
Polyaniline (emeraldine salt), average Mw >15,000, powder (Infusible), 3-100 μm particle size
Pricing and availability is not currently available.