MilliporeSigma
  • Pigment epithelium derived factor as a novel multi-target treatment for uterine fibroids.
Pigment epithelium derived factor as a novel multi-target treatment for uterine fibroids.
Reproductive biomedicine online (2020-06-14)
Hadas Bar-Joseph, Elad Hikri, Dana Chuderland, Ido Ben-Ami, Ruth Shalgi
ABSTRACT
Does recombinant pigment epithelium derived factor (PEDF) have potential in treating uterine fibroids? In-vitro models that used human leiomyoma and Eker rat uterine leiomyoma (ELT-3) cell lines. The ELT-3 cell line was used to examine cellular targets after adding recombinant PEDF to the culture media. Athymic nude female mice were used as an in-vivo model. They were injected with ELT-3 cells to induce ectopic fibroid lesions, then treated with recombinant PEDF. RNA expression of PEDF and its receptors was found in both leiomyoma cell lines, as well as the expression of PEDF receptors. Addition of recombinant PEDF to the culture medium of leiomyoma cell lines activated ERK in a time-dependent manner, induced down-regulation of vascular endothelial growth factor mRNA and protein, as well as the mRNAs of oestrogen receptors alpha and beta and inhibited cellular proliferation. Treatment of mice-bearing fibroids with recombinant PEDF reduced fibroid growth rate and resulted in smaller tumours. This study suggests that recombinant PEDF is a putative novel potent physiological treatment for uterine fibroids. It targets several cornerstones of fibroid pathobiology in parallel, including vascular endothelial growth factor and oestrogen receptors, which are needed for vascularization, and restricts fibroid growth and final size in an animal model.
Sigma-Aldrich
β-Estradiol 17-valerate, ≥98%
Sigma-Aldrich
REDTaq® ReadyMix PCR Reaction Mix, Complete PCR reagent with standard Taq DNA Polymerase and inert dye
Millipore
Ni-NTA His•Bind® Resin
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture