MilliporeSigma
  • Pharmacokinetics and pharmacodynamics of TTI-101, a STAT3 inhibitor that blocks muscle proteolysis in rats with chronic kidney disease.
Pharmacokinetics and pharmacodynamics of TTI-101, a STAT3 inhibitor that blocks muscle proteolysis in rats with chronic kidney disease.
American journal of physiology. Renal physiology (2020-06-02)
Liping Zhang, Ying Wang, Yanlan Dong, Zihong Chen, Thomas K Eckols, Moses M Kasembeli, David J Tweardy, William E Mitch
ABSTRACT
Loss of muscle proteins increases the morbidity and mortality of patients with chronic kidney disease (CKD), and there are no reliable preventive treatments. We uncovered a STAT3/CCAAT-enhancer-binding protein-δ to myostatin signaling pathway that activates muscle protein degradation in mice with CKD or cancer; we also identified a small-molecule inhibitor of STAT3 (TTI-101) that blocks this pathway. To evaluate TTI-101 as a treatment for CKD-induced cachexia, we measured TTI-101 pharmacokinetics and pharmacodynamics in control and CKD rats that were orally administered TTI-101or its diluent. The following two groups of gavage-fed rats were studied: sham-operated control rats and CKD rats. Plasma was collected serially (0, 0.25, 0.5, 1, 2, 4, 8, and 24 h) following TTI-101 administration (at oral doses of 0, 10, 30, or 100 mg/kg). Plasma levels of TTI-101 were measured by LC-MS/MS, and pharmacokinetic results were analyzed with the PKSolver program. Plasma TTI-101 levels increased linearly with doses; the maximum plasma concentrations and time to maximal plasma levels (~1 h) were similar in sham-operated control rats and CKD rats. Notably, gavage treatment of TTI-101 for 3 days produced TTI-101 muscle levels in sham control rats and CKD rats that were not significantly different. CKD rats that received TTI-101 for 7 days had suppression of activated STAT3 and improved muscle grip strength; there also was a trend for increasing body and muscle weights. TTI-101 was tolerated at doses of 100 mg·kg-1·day-1 for 7 days. These results with TTI-101 in rats warrant its development as a treatment for cachexia in humans.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-ACTIN antibody produced in rabbit, affinity isolated antibody