MilliporeSigma
  • Dynamic regulation of Z-DNA in the mouse prefrontal cortex by the RNA-editing enzyme Adar1 is required for fear extinction.
Dynamic regulation of Z-DNA in the mouse prefrontal cortex by the RNA-editing enzyme Adar1 is required for fear extinction.
Nature neuroscience (2020-05-06)
Paul R Marshall, Qiongyi Zhao, Xiang Li, Wei Wei, Ambika Periyakaruppiah, Esmi L Zajaczkowski, Laura J Leighton, Sachithrani U Madugalle, Dean Basic, Ziqi Wang, Jiayu Yin, Wei-Siang Liau, Ankita Gupte, Carl R Walkley, Timothy W Bredy
ABSTRACT
DNA forms conformational states beyond the right-handed double helix; however, the functional relevance of these noncanonical structures in the brain remains unknown. Here we show that, in the prefrontal cortex of mice, the formation of one such structure, Z-DNA, is involved in the regulation of extinction memory. Z-DNA is formed during fear learning and reduced during extinction learning, which is mediated, in part, by a direct interaction between Z-DNA and the RNA-editing enzyme Adar1. Adar1 binds to Z-DNA during fear extinction learning, which leads to a reduction in Z-DNA at sites where Adar1 is recruited. Knockdown of Adar1 leads to an inability to modify a previously acquired fear memory and blocks activity-dependent changes in DNA structure and RNA state-effects that are fully rescued by the introduction of full-length Adar1. These findings suggest a new mechanism of learning-induced gene regulation that is dependent on proteins that recognize alternate DNA structure states, which are required for memory flexibility.
Sigma-Aldrich
Poly-L-ornithine hydrochloride, mol wt 15,000-30,000
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells