MilliporeSigma
Star-Shaped Ruthenium Complexes as Prototypes of Molecular Gears.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-10-12)
Guillaume Erbland, Seifallah Abid, Yohan Gisbert, Nathalie Saffon-Merceron, Yuichiro Hashimoto, Leonardo Andreoni, Théo Guérin, Claire Kammerer, Gwénaël Rapenne
ABSTRACT
The design and synthesis of two families of molecular-gear prototypes is reported, with the aim of assembling them into trains of gears on a surface and ultimately achieving controlled intermolecular gearing motion. These piano-stool ruthenium complexes incorporate a hydrotris(indazolyl)borate moiety as tripodal rotation axle and a pentaarylcyclopentadienyl ligand as star-shaped cogwheel, equipped with five teeth ranging from pseudo-1D aryl groups to large planar 2D paddles. A divergent synthetic approach was followed, starting from a pentakis(p-bromophenyl)cyclopentadienyl ruthenium(II) complex as key precursor or from its iodinated counterpart, obtained by copper-catalyzed aromatic Br/I exchange. Subsequent fivefold cross-coupling reactions with various partners allowed high structural diversity to be reached and yielded molecular-gear prototypes with aryl-, carbazole-, BODIPY- and porphyrin-derived teeth of increasing size and length.
Sigma-Aldrich
4-tert-Butylphenylboronic acid, ≥95.0%
Sigma-Aldrich
3-Ethyl-2,4-dimethylpyrrole, 97%
Sigma-Aldrich
3,6-Di-tert-butylcarbazole, 97%