MilliporeSigma
  • Cellular sensing of extracellular purine nucleosides triggers an innate IFN-β response.
Cellular sensing of extracellular purine nucleosides triggers an innate IFN-β response.
Science advances (2020-08-04)
Rekha Dhanwani, Mariko Takahashi, Ian T Mathews, Camille Lenzi, Artem Romanov, Jeramie D Watrous, Bartijn Pieters, Catherine C Hedrick, Chris A Benedict, Joel Linden, Roland Nilsson, Mohit Jain, Sonia Sharma
ABSTRACT
Mechanisms linking immune sensing of DNA danger signals in the extracellular environment to innate pathways in the cytosol are poorly understood. Here, we identify a previously unidentified immune-metabolic axis by which cells respond to purine nucleosides and trigger a type I interferon-β (IFN-β) response. We find that depletion of ADA2, an ectoenzyme that catabolizes extracellular dAdo to dIno, or supplementation of dAdo or dIno stimulates IFN-β. Under conditions of reduced ADA2 enzyme activity, dAdo is transported into cells and undergoes catabolysis by the cytosolic isoenzyme ADA1, driving intracellular accumulation of dIno. dIno is a functional immunometabolite that interferes with the cellular methionine cycle by inhibiting SAM synthetase activity. Inhibition of SAM-dependent transmethylation drives epigenomic hypomethylation and overexpression of immune-stimulatory endogenous retroviral elements that engage cytosolic dsRNA sensors and induce IFN-β. We uncovered a previously unknown cellular signaling pathway that responds to extracellular DNA-derived metabolites, coupling nucleoside catabolism by adenosine deaminases to cellular IFN-β production.
Sigma-Aldrich
Anti-CECR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Bis(diphenylphosphino)methane, 97%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Cycloleucine, 97%
Sigma-Aldrich
Poly(deoxyadenylic-thymidylic) acid sodium salt, double-stranded alternating copolymer
Sigma-Aldrich
Inosine, ≥99% (HPLC)