MilliporeSigma
  • The effects of feeding a high-fiber or high-starch pellet at two daily allocations on feed intake patterns, rumen fermentation, and milk production of mid-lactation dairy cows.
The effects of feeding a high-fiber or high-starch pellet at two daily allocations on feed intake patterns, rumen fermentation, and milk production of mid-lactation dairy cows.
Journal of dairy science (2020-04-21)
J Haisan, M Oba
ABSTRACT
The objective of this experiment was to determine the effect of pellet type and feeding amount on feeding behavior, dry matter intake, rumen fermentation, and milk production of lactating dairy cows. An experimental diet was formulated to provide an adequate amount of nutrients to a 650-kg cow producing 40 kg of milk per day, with a portion of the diet removed as a high-fiber (33.2% neutral detergent fiber; F) or high-starch (56.8% starch; S) pellet. Pellets were fed at a low (1 kg; L) or high (3 kg; H) amount twice per day alongside a partial mixed ration (PMR). Four complementary PMR were formulated for each pellet treatment such that the overall diet (pellet + PMR) offered to the cows was the same among all treatments. Eight ruminally cannulated cows were used in a 4 × 4 Latin square design with 14-d periods. Cows were fed PMR once daily at 1200 h, and pellet twice daily at 0600 and 1800 h. Data and samples were collected on d 11 to 14 of each period. By design there was a difference in pellet intake between the H and L treatments (5.31 vs. 1.81 kg/d), and PMR intake was reduced when H pellet was fed (22.9 vs. 25.3 kg/d); however, feeding H tended to increase total dry matter intake. Feed disappearance, which was measured as the amount of PMR consumed every 3 h following PMR delivery, was affected by the nutrient composition of the PMR as cows fed S (with high-fiber PMR) consumed 28.6% of their PMR intake within 3 h of delivery, whereas cows fed F (with high-starch PMR) consumed 33.5%. Duration that pH was below 5.8 tended to be lower when cows were fed the S pellet (270 vs. 125 min/d) compared with F. In addition, feeding the S pellet (with high-fiber PMR) decreased plasma concentrations of glucose (66.0 vs. 70.0 mg/dL) and insulin (1.90 vs. 2.25 ng/mL) compared with F. These results suggest that the composition of the PMR dictates rumen fermentation to a greater extent than composition of pellets. The S pellet was fed alongside a high-fiber PMR, which was more filling in the rumen, less fermentable, and contained more neutral detergent fiber. Although no difference was observed in milk production among treatments, the fact that feed intake pattern and rumen fermentation are better explained by nutrient composition of the PMR should be considered when formulating diets for lactating cows fed pellet and PMR, such as those milked with automated milking systems.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
PGO Enzyme Preparation, 1 G capsules