MilliporeSigma
  • Topoisomerase 3β knockout mice show transcriptional and behavioural impairments associated with neurogenesis and synaptic plasticity.
Topoisomerase 3β knockout mice show transcriptional and behavioural impairments associated with neurogenesis and synaptic plasticity.
Nature communications (2020-06-21)
Yuyoung Joo, Yutong Xue, Yue Wang, Ross A McDevitt, Nirnath Sah, Simone Bossi, Shuaikun Su, Seung Kyu Lee, Wei Peng, Aoji Xie, Yongqing Zhang, Yi Ding, Wai Lim Ku, Soumita Ghosh, Kenneth Fishbein, Weiping Shen, Richard Spencer, Kevin Becker, Keji Zhao, Mark P Mattson, Henriette van Praag, Alexei Sharov, Weidong Wang
ABSTRACT
Topoisomerase 3β (Top3β) is the only dual-activity topoisomerase in animals that can change topology for both DNA and RNA, and facilitate transcription on DNA and translation on mRNAs. Top3β mutations have been linked to schizophrenia, autism, epilepsy, and cognitive impairment. Here we show that Top3β knockout mice exhibit behavioural phenotypes related to psychiatric disorders and cognitive impairment. The mice also display impairments in hippocampal neurogenesis and synaptic plasticity. Notably, the brains of the mutant mice exhibit impaired global neuronal activity-dependent transcription in response to fear conditioning stress, and the affected genes include many with known neuronal functions. Our data suggest that Top3β is essential for normal brain function, and that defective neuronal activity-dependent transcription may be a mechanism by which Top3β deletion causes cognitive impairment and psychiatric disorders.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-TOP3B antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution