MilliporeSigma
  • Preventive effects of folic acid on Zika virus-associated poor pregnancy outcomes in immunocompromised mice.
Preventive effects of folic acid on Zika virus-associated poor pregnancy outcomes in immunocompromised mice.
PLoS pathogens (2020-05-12)
Yogy Simanjuntak, Hui-Ying Ko, Yi-Ling Lee, Guann-Yi Yu, Yi-Ling Lin
ABSTRACT
Zika virus (ZIKV) infection may lead to congenital microcephaly and pregnancy loss in pregnant women. In the context of pregnancy, folic acid (FA) supplementation may reduce the risk of abnormal pregnancy outcomes. Intriguingly, FA may have a beneficial effect on the adverse pregnancy outcomes associated with ZIKV infection. Here, we show that FA inhibits ZIKV replication in human umbilical vein endothelial cells (HUVECs) and a cell culture model of blood-placental barrier (BPB). The inhibitory effect of FA against ZIKV infection is associated with FRα-AMPK signaling. Furthermore, treatment with FA reduces pathological features in the placenta, number of fetal resorptions, and stillbirths in two mouse models of in utero ZIKV transmission. Mice with FA treatment showed lower viral burden and better prognostic profiles in the placenta including reduced inflammatory response, and enhanced integrity of BPB. Overall, our findings suggest the preventive role of FA supplementation in ZIKV-associated abnormal pregnancy and warrant nutritional surveillance to evaluate maternal FA status in areas with active ZIKV transmission.
Sigma-Aldrich
Endothelial cell growth supplement from bovine neural tissue, ECGS, suitable for cell culture
Sigma-Aldrich
Ebselen, cysteine modifier
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Bouin′s solution, histological fixative
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 70,000